4-Chloro-3-methylphenyl-3-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050727-500MG 050727-2.5G
101892-808EA 364.62 USD
101892-808 101892-810
4-Chloro-3-methylphenyl-3-piperidinylmethyl ether hydrochloride
4-Chloro-3-methylphenyl-3-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MW: 276.21 g/mol
MDL Number: MFCD13561115
CAS Number: 1220018-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR