3-(4-Chloro-3-methylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050725-500MG 050725-2.5G
101892-800EA 364.62 USD
101892-800 101892-802
3-(4-Chloro-3-methylphenoxy)piperidine hydrochloride
3-(4-Chloro-3-methylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13561113
CAS Number: 1220029-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR