3-(2-Bromo-4-chlorophenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050724-500MG 050724-2.5G
101892-796EA 364.62 USD
101892-796 101892-798
3-(2-Bromo-4-chlorophenoxy)pyrrolidine hydrochloride
3-(2-Bromo-4-chlorophenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂BrCl₂NO
MDL Number: MFCD13561112
CAS Number: 1220033-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR