4-[(2-Bromo-4-chlorophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050723-500MG 050723-2.5G
101892-792EA 364.62 USD
101892-792 101892-794
4-[(2-Bromo-4-chlorophenoxy)methyl]piperidine hydrochloride
4-[(2-Bromo-4-chlorophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆BrCl₂NO
MDL Number: MFCD13561111
CAS Number: 1219972-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR