3-[(2-Bromo-4-chlorophenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050722-500MG 050722-2.5G
101892-788EA 364.62 USD
101892-788 101892-790
3-[(2-Bromo-4-chlorophenoxy)methyl]piperidine hydrochloride
3-[(2-Bromo-4-chlorophenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆BrCl₂NO
MDL Number: MFCD13561110
CAS Number: 1220028-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR