4-(2-Bromo-4-chlorophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050721-500MG 050721-2.5G
101892-784EA 186.15 USD
101892-784 101892-786
4-(2-Bromo-4-chlorophenoxy)piperidine hydrochloride
4-(2-Bromo-4-chlorophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄BrCl₂NO
MDL Number: MFCD13561109
CAS Number: 1220018-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR