3-(2-Bromo-4-chlorophenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050720-500MG 050720-2.5G
101892-780EA 364.62 USD
101892-780 101892-782
3-(2-Bromo-4-chlorophenoxy)piperidine hydrochloride
3-(2-Bromo-4-chlorophenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄BrCl₂NO
MDL Number: MFCD13561108
CAS Number: 1220028-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR