4-(4-Bromophenoxy)piperidine hydrochloride
Supplier: Matrix Scientific
050719-500MG
050719-2.5G
101892-776EA
221.54
USD
101892-776
101892-778
4-(4-Bromophenoxy)piperidine hydrochloride
4-(4-Bromophenoxy)piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₅BrClNO |
MDL Number:
MFCD08059305 CAS Number: 63843-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...