3-(3,4-Dimethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050718-500MG 050718-2.5G
101892-772EA 461.54 USD
101892-772 101892-774
3-(3,4-Dimethylphenoxy)pyrrolidine hydrochloride
3-(3,4-Dimethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClNO
MDL Number: MFCD09998966
CAS Number: 1220033-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR