4-[(3,4-Dimethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050717-500MG 050717-2.5G
101892-768EA 364.62 USD
101892-768 101892-770
4-[(3,4-Dimethylphenoxy)methyl]piperidine hydrochloride
4-[(3,4-Dimethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13561107
CAS Number: 1220028-16-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR