4-(3,4-Dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050715-500MG 050715-2.5G
101892-760EA 166.15 USD
101892-760 101892-762
4-(3,4-Dimethylphenoxy)piperidine hydrochloride
4-(3,4-Dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD11858065
CAS Number: 1170242-76-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR