3-(3,4-Dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050714-500MG 050714-2.5G
101892-756EA 364.62 USD
101892-756 101892-758
3-(3,4-Dimethylphenoxy)piperidine hydrochloride
3-(3,4-Dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD13561105
CAS Number: 1220033-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR