3-[3-(tert-Butyl)phenoxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050713-500MG 050713-2.5G
101892-752EA 364.62 USD
101892-752 101892-754
3-[3-(tert-Butyl)phenoxy]pyrrolidine hydrochloride
3-[3-(tert-Butyl)phenoxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₂ClNO
MDL Number: MFCD13561104
CAS Number: 1220037-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR