3-(tert-Butyl)phenyl-3-piperidinylmethyl ether hydrochloride
Supplier: Matrix Scientific
050711-500MG
050711-2.5G
101892-744EA
364.62
USD
101892-744
101892-746
3-(tert-Butyl)phenyl-3-piperidinylmethyl ether hydrochloride
3-(tert-Butyl)phenyl-3-piperidinylmethyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₆H₂₆ClNO |
MDL Number:
MFCD13561102 CAS Number: 1220028-76-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...