3-(4-Chloro-2-ethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050481-500MG 050481-2.5G
101891-824EA 364.62 USD
101891-824 101891-826
3-(4-Chloro-2-ethylphenoxy)pyrrolidine hydrochloride
3-(4-Chloro-2-ethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560882
CAS Number: 1220028-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR