4-[(4-Chloro-2-ethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050480-500MG 050480-2.5G
101891-820EA 364.62 USD
101891-820 101891-822
4-[(4-Chloro-2-ethylphenoxy)methyl]piperidine hydrochloride
4-[(4-Chloro-2-ethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560881
CAS Number: 1220019-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR