3-[(4-Chloro-2-ethylphenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050479-500MG
050479-2.5G
101891-816EA
364.62
USD
101891-816
101891-818
3-[(4-Chloro-2-ethylphenoxy)methyl]piperidine hydrochloride
3-[(4-Chloro-2-ethylphenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₄H₂₁Cl₂NO |
MDL Number:
MFCD13560880 CAS Number: 1220028-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...