4-(4-Chloro-2-ethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050478-500MG 050478-2.5G
101891-812EA 364.62 USD
101891-812 101891-814
4-(4-Chloro-2-ethylphenoxy)piperidine hydrochloride
4-(4-Chloro-2-ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560879
CAS Number: 1220030-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR