3-(4-Chloro-2-ethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050477-500MG 050477-2.5G
101891-808EA 364.62 USD
101891-808 101891-810
3-(4-Chloro-2-ethylphenoxy)piperidine hydrochloride
3-(4-Chloro-2-ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560878
CAS Number: 1220018-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR