4-[(2-Chloro-4-methoxyphenoxy)methyl]piperidine hydrochloride
Supplier: Matrix Scientific
050475-500MG
050475-2.5G
101891-800EA
364.62
USD
101891-800
101891-802
4-[(2-Chloro-4-methoxyphenoxy)methyl]piperidine hydrochloride
4-[(2-Chloro-4-methoxyphenoxy)methyl]piperidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉Cl₂NO₂ |
MDL Number:
MFCD13560876 CAS Number: 1220035-87-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...