3-[(2-Chloro-4-methoxyphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050474-500MG 050474-2.5G
101891-796EA 364.62 USD
101891-796 101891-798
3-[(2-Chloro-4-methoxyphenoxy)methyl]piperidine hydrochloride
3-[(2-Chloro-4-methoxyphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO₂
MDL Number: MFCD13560875
CAS Number: 1220020-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR