2-Chloro-4-methoxyphenyl-3-piperidinyl ether hydrochloride
Supplier: Matrix Scientific
050472-500MG
050472-2.5G
101891-788EA
364.62
USD
101891-788
101891-790
2-Chloro-4-methoxyphenyl-3-piperidinyl ether hydrochloride
2-Chloro-4-methoxyphenyl-3-piperidinyl ether hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂NO₂ |
MDL Number:
MFCD13560873 CAS Number: 1220018-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...