3-(4-Chloro-2-isopropylphenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
050471-500MG
050471-2.5G
101891-784EA
166.15
USD
101891-784
101891-786
3-(4-Chloro-2-isopropylphenoxy)pyrrolidine hydrochloride
3-(4-Chloro-2-isopropylphenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉Cl₂NO |
MDL Number:
MFCD13560872 CAS Number: 1220027-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...