4-[(4-Chloro-2-isopropylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050470-500MG 050470-2.5G
101891-780EA 364.62 USD
101891-780 101891-782
4-[(4-Chloro-2-isopropylphenoxy)methyl]piperidine hydrochloride
4-[(4-Chloro-2-isopropylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560871
CAS Number: 1220032-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR