4-(4-Chloro-2-isopropylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050468-500MG 050468-2.5G
101891-772EA 364.62 USD
101891-772 101891-774
4-(4-Chloro-2-isopropylphenoxy)piperidine hydrochloride
4-(4-Chloro-2-isopropylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560869
CAS Number: 1220020-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR