3-(2-Chloro-4-ethylphenoxy)pyrrolidine hydrochloride
Supplier: Matrix Scientific
050466-500MG
050466-2.5G
101891-764EA
364.62
USD
101891-764
101891-766
3-(2-Chloro-4-ethylphenoxy)pyrrolidine hydrochloride
3-(2-Chloro-4-ethylphenoxy)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂NO |
MDL Number:
MFCD13560867 CAS Number: 1219977-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...