3-[(2-Chloro-4-ethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050464-500MG 050464-2.5G
101891-756EA 364.62 USD
101891-756 101891-758
3-[(2-Chloro-4-ethylphenoxy)methyl]piperidine hydrochloride
3-[(2-Chloro-4-ethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560865
CAS Number: 1220035-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR