4-(2-Chloro-4-ethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050463-500MG 050463-2.5G
101891-752EA 364.62 USD
101891-752 101891-754
4-(2-Chloro-4-ethylphenoxy)piperidine hydrochloride
4-(2-Chloro-4-ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560864
CAS Number: 1220020-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR