3-(2-Chloro-4-ethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050462-500MG 050462-2.5G
101891-748EA 364.62 USD
101891-748 101891-750
3-(2-Chloro-4-ethylphenoxy)piperidine hydrochloride
3-(2-Chloro-4-ethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560863
CAS Number: 1220018-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR