3-(2-Chloro-4-methylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050461-500MG 050461-2.5G
101891-744EA 364.62 USD
101891-744 101891-746
3-(2-Chloro-4-methylphenoxy)pyrrolidine hydrochloride
3-(2-Chloro-4-methylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅Cl₂NO
MDL Number: MFCD13560862
CAS Number: 1219977-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR