2-Chloro-4-methylphenyl-3-piperidinylmethyl ether hydrochloride

Supplier: Matrix Scientific

050459-500MG 050459-2.5G
101891-736EA 364.62 USD
101891-736 101891-738
2-Chloro-4-methylphenyl-3-piperidinylmethyl ether hydrochloride
2-Chloro-4-methylphenyl-3-piperidinylmethyl ether hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560860
CAS Number: 1220027-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR