3-(2-Chloro-4-methylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050457-500MG 050457-2.5G
101891-728EA 364.62 USD
101891-728 101891-730
3-(2-Chloro-4-methylphenoxy)piperidine hydrochloride
3-(2-Chloro-4-methylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MW: 262.18 g/mol
MDL Number: MFCD13560858
CAS Number: 1220035-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR