3-[4-(sec-Butyl)-2-chlorophenoxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050456-500MG 050456-2.5G
101891-724EA 166.15 USD
101891-724 101891-726
3-[4-(sec-Butyl)-2-chlorophenoxy]pyrrolidine hydrochloride
3-[4-(sec-Butyl)-2-chlorophenoxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560857
CAS Number: 1220027-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR