4-[4-(sec-Butyl)-2-chlorophenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050453-500MG 050453-2.5G
101891-712EA 364.62 USD
101891-712 101891-714
4-[4-(sec-Butyl)-2-chlorophenoxy]piperidine hydrochloride
4-[4-(sec-Butyl)-2-chlorophenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560854
CAS Number: 1220030-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR