3-[4-(sec-Butyl)-2-chlorophenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050452-500MG 050452-2.5G
101891-708EA 364.62 USD
101891-708 101891-710
3-[4-(sec-Butyl)-2-chlorophenoxy]piperidine hydrochloride
3-[4-(sec-Butyl)-2-chlorophenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560853
CAS Number: 1220019-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR