3-(2-Chloro-4-isopropylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050451-500MG 050451-2.5G
101891-704EA 364.62 USD
101891-704 101891-706
3-(2-Chloro-4-isopropylphenoxy)pyrrolidine hydrochloride
3-(2-Chloro-4-isopropylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560852
CAS Number: 1220018-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR