4-[(2-Chloro-4-isopropylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050450-500MG 050450-2.5G
101891-700EA 364.62 USD
101891-700 101891-702
4-[(2-Chloro-4-isopropylphenoxy)methyl]piperidine hydrochloride
4-[(2-Chloro-4-isopropylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560851
CAS Number: 1220019-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR