3-[(2-Chloro-4-isopropylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050449-500MG 050449-2.5G
101891-696EA 364.62 USD
101891-696 101891-698
3-[(2-Chloro-4-isopropylphenoxy)methyl]piperidine hydrochloride
3-[(2-Chloro-4-isopropylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₅H₂₃Cl₂NO
MDL Number: MFCD13560850
CAS Number: 1220035-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR