3-(2-Chloro-4-isopropylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050447-500MG 050447-2.5G
101891-688EA 364.62 USD
101891-688 101891-690
3-(2-Chloro-4-isopropylphenoxy)piperidine hydrochloride
3-(2-Chloro-4-isopropylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560848
CAS Number: 1220035-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR