3-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]pyrrolidine hydrochloride

Supplier: Matrix Scientific

050446-500MG 050446-2.5G
101891-684EA 364.62 USD
101891-684 101891-686
3-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]pyrrolidine hydrochloride
3-[(3-Chloro[1,1'-biphenyl]-4-yl)oxy]pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₁₇Cl₂NO
MDL Number: MFCD13560847
CAS Number: 1219976-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR