3-Chloro[1,1'-biphenyl]-4-yl-4-piperidinylmethylether hydrochloride

Supplier: Matrix Scientific

050445-500MG 050445-2.5G
101891-680EA 364.62 USD
101891-680 101891-682
3-Chloro[1,1'-biphenyl]-4-yl-4-piperidinylmethylether hydrochloride
3-Chloro[1,1'-biphenyl]-4-yl-4-piperidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁Cl₂NO
MW: 338.28 g/mol
MDL Number: MFCD13560846
CAS Number: 1219976-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR