3-(2-Chloro-4,6-dimethylphenoxy)pyrrolidine hydrochloride

Supplier: Matrix Scientific

050441-500MG 050441-2.5G
101891-664EA 364.62 USD
101891-664 101891-666
3-(2-Chloro-4,6-dimethylphenoxy)pyrrolidine hydrochloride
3-(2-Chloro-4,6-dimethylphenoxy)pyrrolidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇Cl₂NO
MDL Number: MFCD13560842
CAS Number: 1220035-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR