4-[(2-Chloro-4,6-dimethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050440-500MG 050440-2.5G
101891-660EA 364.62 USD
101891-660 101891-662
4-[(2-Chloro-4,6-dimethylphenoxy)methyl]piperidine hydrochloride
4-[(2-Chloro-4,6-dimethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560841
CAS Number: 1219971-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR