3-[(2-Chloro-4,6-dimethylphenoxy)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

050439-500MG 050439-2.5G
101891-656EA 364.62 USD
101891-656 101891-658
3-[(2-Chloro-4,6-dimethylphenoxy)methyl]piperidine hydrochloride
3-[(2-Chloro-4,6-dimethylphenoxy)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁Cl₂NO
MDL Number: MFCD13560840
CAS Number: 1220035-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR