4-(2-Chloro-4,6-dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050438-500MG 050438-2.5G
101891-652EA 364.62 USD
101891-652 101891-654
4-(2-Chloro-4,6-dimethylphenoxy)piperidine hydrochloride
4-(2-Chloro-4,6-dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560839
CAS Number: 1220027-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR