3-(2-Chloro-4,6-dimethylphenoxy)piperidine hydrochloride

Supplier: Matrix Scientific

050437-500MG 050437-2.5G
101891-648EA 364.62 USD
101891-648 101891-650
3-(2-Chloro-4,6-dimethylphenoxy)piperidine hydrochloride
3-(2-Chloro-4,6-dimethylphenoxy)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉Cl₂NO
MDL Number: MFCD13560838
CAS Number: 1220019-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR