4-{[4-(tert-Butyl)-2-methylphenoxy]methyl}piperidine hydrochloride

Supplier: Matrix Scientific

050435-500MG 050435-2.5G
101891-640EA 364.62 USD
101891-640 101891-642
4-{[4-(tert-Butyl)-2-methylphenoxy]methyl}piperidine hydrochloride
4-{[4-(tert-Butyl)-2-methylphenoxy]methyl}piperidine hydrochloride

Hazard: Irritant

Formula: C₁₇H₂₈ClNO
MDL Number: MFCD13560836
CAS Number: 1220018-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR