3-[4-(tert-Butyl)-2-methylphenoxy]piperidine hydrochloride

Supplier: Matrix Scientific

050432-500MG 050432-2.5G
101891-628EA 364.62 USD
101891-628 101891-630
3-[4-(tert-Butyl)-2-methylphenoxy]piperidine hydrochloride
3-[4-(tert-Butyl)-2-methylphenoxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆ClNO
MDL Number: MFCD13560833
CAS Number: 1220030-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR