5-Bromo[1,1'-biphenyl]-2-yl-3-piperidinylmethylether hydrochloride

Supplier: Matrix Scientific

050429-500MG 050429-2.5G
101891-616EA 364.62 USD
101891-616 101891-618
5-Bromo[1,1'-biphenyl]-2-yl-3-piperidinylmethylether hydrochloride
5-Bromo[1,1'-biphenyl]-2-yl-3-piperidinylmethylether hydrochloride

Hazard: Irritant

Formula: C₁₈H₂₁BrClNO
MW: 382.74 g/mol
MDL Number: MFCD13560830
CAS Number: 1220032-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR