5-Bromo[1,1'-biphenyl]-2-yl-4-piperidinyl ether hydrochloride

Supplier: Matrix Scientific

050428-500MG 050428-2.5G
101891-612EA 364.62 USD
101891-612 101891-614
5-Bromo[1,1'-biphenyl]-2-yl-4-piperidinyl ether hydrochloride
5-Bromo[1,1'-biphenyl]-2-yl-4-piperidinyl ether hydrochloride

Hazard: Irritant

Formula: C₁₇H₁₉BrClNO
MDL Number: MFCD13560829
CAS Number: 1220027-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR